missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MSP/MST1 Recombinant Protein

Brand:  R&D Systems™ 352-MS-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463144

  • £904.91 / 50µg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Measured by its ability to activate MSP R/Ron in MDA-MB-453 or T47D human breast cancer cells. 80 ng/mL of the rhMSP significantly induces phosphorylation of MSP R/Ron measured by the Human Phospho-MSP R/Ron DuoSet ® IC Kit (Catalog # DYC1947 ).
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
78.5 kDa (MSP alpha/beta); 53.3 kDa ( alpha chain), 25.2 kDa (beta chain)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer
Yes
4485
MSP/MST1
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos