missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MSP/MST1 β Chain Recombinant Protein

Brand:  R&D Systems™ 635-MS-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465774

  • £316.81 / 25µg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Val484-Gly711
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 4485
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
25 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
4485
MSP/MST1 β Chain
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos