missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse Wnt-5a Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 645-WN-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465814

  • £421.89 / 10µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human/Mouse Wnt-5a Protein is derived from CHO. The Recombinant Human/Mouse Wnt-5a Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
38 kDa
80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human/Mouse
Unlabeled
7474
Wnt-5a
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents