missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse/Rat Activin A Recombinant Protein

For use in cell or tissue culture applications.

Brand:  R&D Systems™ 338-AC-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462934

  • £287.77 / 10µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

  • Source: Chinese Hamster Ovary cell line, CHO-derived, Gly311-Ser426
  • Purity: 95%, by SDS-PAGE with silver staining.
  • Molecular Weight: 13 kDa (monomer)
  • Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
  • Entrez Gene ID: 3624
  • Regulatory Status: RUO
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
13 kDa (monomer)
95%, by SDS-PAGE with silver staining.
Human/Mouse/Rat
Unlabeled
3624
Activin A
10μg
Disulfide-linked homodimer
Yes
Videos