missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse Indian Hedgehog/Ihh (C28II), N-Terminus Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1705-HH-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438673

  • £368.29 / 25µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human/Mouse Indian Hedgehog (C28II) N-Term is derived from E. coli. The Recombinant Human/Mouse Indian Hedgehog (C28II) N-Term has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose.
20 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human/Mouse
Unlabeled
3549
Indian Hedgehog/Ihh (C28II), N-Terminus
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos