missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Mouse Indian Hedgehog/Ihh (C28II), N-Terminus Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 3549 |
Molecular Weight (g/mol) | 20 kDa |
Name | Indian Hedgehog/Ihh (C28II), N-Terminus |
Quantity | 25μg |
Description
The Recombinant Human/Mouse Indian Hedgehog/Ihh (C28II) N-Term is derived from E. coli. The Recombinant Human/Mouse Indian Hedgehog/Ihh (C28II) N-Term has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
20 kDa | |
25μg | |
E. coli-derived Indian Hedgehog/Ihh protein Cys28-Gly202 (Cys28Ile-Ile),with an N-terminal Met |
3549 | |
Indian Hedgehog/Ihh (C28II), N-Terminus | |
Yes |