missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Mouse FGF-8 b Isoform Recombinant Protein

Brand:  R&D Systems™ 423-F8-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464414

  • £294.37 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Gln23-Arg215, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS and Na2SO4.
Entrez Gene ID: 14179
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and Brij-35.
22.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human/Mouse
Yes
14179
FGF-8 b Isoform
25μg
Unlabeled
Videos