missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human M-CSF Recombinant Protein, (CHO-expressed)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 1435 |
Molecular Weight (g/mol) | 25.1 kDa (monomer) |
Name | M-CSF |
Quantity | 10μg |
Description
The Recombinant Human M-CSF (CHO-expressed) Protein is derived from CHO. The Recombinant Human M-CSF (CHO-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
25.1 kDa (monomer) | |
10μg | |
Chinese Hamster Ovary cell line,CHO-derived human M-CSF protein Glu33-Arg255 |
1435 | |
M-CSF | |
Yes |