missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human M-CSF Recombinant Protein, (CHO-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 216-MCC-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13430714

  • £332.22 / 10µg

Estimated Shipment date
from Supplier 21-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human M-CSF (CHO-expressed) Protein is derived from CHO. The Recombinant Human M-CSF (CHO-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
25.1 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked homodimer
Yes
1435
M-CSF
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents