missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Lymphotoxin α2/β1 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 679-TX-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13435944

  • £364.33 / 10µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Lymphotoxin alpha2/beta1 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Lymphotoxin alpha2/beta1 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
18 kDa (LT alpha), 18.5 kDa ( LTbeta)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Noncovalently-linked heterotrimer
Yes
4049
Lymphotoxin α2/β1
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos