missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human LIF Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 7734-LF-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13446294

  • £242.55 / 25µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human LIF Protein is derived from E. coli. The Recombinant Human LIF Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
19.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Yes
3976
LIF
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents