missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Leptin R Fc Chimera Recombinant Protein

Brand:  R&D Systems™ 389-LR-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463784

  • £260.05 / 100µg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Measured by its ability to inhibit Leptin-dependent proliferation of BaF3 mouse pro-B cells transfected with human Leptin R. The ED 50 for this effect is 0.02-0.12 µg/mL in the presence of 3 ng/mL Recombinant Human Leptin/OB (Catalog # 398-LP ) .
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS with BSA as a carrier protein.
121 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Disulfide-linked homodimer
Unlabeled
Reconstitute at 200μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
3953
Leptin R Fc Chimera
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos