missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Leptin/OB Recombinant Protein

Brand:  R&D Systems™ 398-LP-01M

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463934

  • £231.00 / 1mg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Val22-Cys167, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 3952
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
16 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3952
Leptin/OB
1mg
Unlabeled
Videos