missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Layilin Recombinant Protein

Brand:  R&D Systems™ 2536-LA-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461424

  • £298.33 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala22-Glu220, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 143903
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
Observed Molecular Wt: 30-35kDa, reducing conditions, Predicted Molecular Wt.: 24.1kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
143903
Layilin
25μg
Unlabeled
Videos