missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Latent TGF-beta 1 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 299-LT-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13432214

  • £376.32 / 5µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Latent TGF-beta 1 Protein is derived from CHO. The Recombinant Human Latent TGF-beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein.
Observed Molecular Wt: 40-43kDa & 12kDa, reducing conditions 80-90kDa, non-reducing conditions, Predicted Molecular Wt.: 41.3kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Leu30 & Ala279
CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Yes
7040
Latent TGF-beta 1
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, ≤ -20° C under sterile conditions after opening.
<1.0 EU per 1μg of the protein by the LAL method.
Unlabeled
Videos
Documents