missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Latent TGF-beta 1 Recombinant Protein

Brand:  R&D Systems™ 299-LT-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462214

  • £853.72 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Leu30-Ser390
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 41.3 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in PBS and Glycerol.
Entrez Gene ID: 7040
Regulatory Status: RUO

Specifications

Specifications

Supplied as a 0.2μm filtered solution in PBS and Glycerol.
Observed Molecular Wt: 40-43kDa & 12kDa, reducing conditions 80-90kDa, non-reducing conditions, Predicted Molecular Wt.: 41.3kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
<1.0 EU per 1μg of the protein by the LAL method.
Yes
7040
Latent TGF-beta 1
25μg
Leu30 & Ala279
Unlabeled
Videos