missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Latent TGF-beta 1 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 299-LT-005/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13442214

  • £328.69 / 5µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Latent TGF-beta 1 Protein is derived from CHO. The Recombinant Human Latent TGF-beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in PBS and Glycerol.
Observed Molecular Wt: 40-43kDa & 12kDa, reducing conditions 80-90kDa, non-reducing conditions, Predicted Molecular Wt.: 41.3kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Leu30 & Ala279
Unlabeled
7040
Latent TGF-beta 1
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, ≤ -20° C under sterile conditions after opening.
<1.0 EU per 1μg of the protein by the LAL method.
Yes
Videos