missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Latent TGF-beta 1 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
POA
Specifications
Product Type | Human Latent TGF-beta 1 ELISpot Kit |
---|---|
For Use With (Application) | Diagnostics |
Sample Type | Whole Cells |
Description
ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the detection and enumeration of . Complete ELISpot kits are ready-to-run and require no assay development or refinement.
Product Features:- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- Hig
Specifications
Human Latent TGF-beta 1 ELISpot Kit | |
Diagnostics | |
Whole Cells |