missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human KGF/FGF-7 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 2252 |
Molecular Weight (g/mol) | 19 kDa |
Name | KGF/FGF-7 |
Recombinant | Yes |
Description
The Recombinant Human KGF/FGF-7 Protein is derived from E. coli. The Recombinant Human KGF/FGF-7 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
19 kDa | |
Yes |
2252 | |
KGF/FGF-7 | |
E. coli-derived human KGF/FGF-7 protein Cys32-Thr194,with an N-terminal Met |