missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-9 Recombinant Protein

Brand:  R&D Systems™ 209-IL-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460474

  • £319.45 / 10µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gln19-Ile144
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3578
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
14.1 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3578
IL-9
10μg
Unlabeled
Videos