missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-6 Recombinant Protein

Brand:  R&D Systems™ 206-IL-200

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15348884

  • £1040.59 / Each


Estimated Shipment
25-01-2019
Add to basket

Description

Description

Source: E. coli-derived, Pro29-Met212
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 3569
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 100-200μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
3569
IL-6
200μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
20.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
Videos