missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-6 ELISpot Kit

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Brand:  R&D Systems™ EL206

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455095

  • £410.13 / 1 set


Estimated Shipment date
from Supplier 27-03-2020
Add to basket

Description

Description

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • Hig
Specifications

Specifications

Diagnostics
Human IL-6 ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
B cell stimulatory factor-2, B-cell differentiation factor, BSF2, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
One 96-well PVDF-backed plate coated w/polyclonal Ab for human IL-6, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control
Whole Cells
one 96-well plate
Videos
Documents