missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-5 Recombinant Protein

Brand:  R&D Systems™ 205-IL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459423

  • £564.22 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Ile20-Ser134
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.1 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3567
Regulatory Status: RUO

Specifications

Specifications

Supplied as a 0.2μm filtered solution in PBS.
13.1 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3567
IL-5
25μg
Disulfide-linked homodimer
Yes
Videos