missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-5 R alpha/CD125 Recombinant Protein, (CHO-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 7404-5R-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436174

  • £385.45 / 25µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IL-5 R alpha (CHO-expressed) Protein is derived from CHO. The Recombinant Human IL-5 R alpha (CHO-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
35.8 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Monomer
Yes
3568
IL-5 R alpha/CD125
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos