missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-5 R alpha/CD125 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 3568 |
Molecular Weight (g/mol) | 38 kDa |
Name | IL-5R alpha/CD125 |
Quantity | 25μg |
Description
The Recombinant Human IL-5 R alpha Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-5 R alpha Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
38 kDa | |
25μg | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-5 R alpha/CD125 protein Asp21-Glu335 |
3568 | |
IL-5R alpha/CD125 | |
Yes |