missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-36 gamma/IL-1F9 Recombinant Protein

Brand:  R&D Systems™ 2320-IL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461024

  • £245.53 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Met1-Asp169
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 56300
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
18.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
56300
IL-36 gamma/IL-1F9
25μg
Unlabeled
Videos