missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-33 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 90865 |
Molecular Weight (g/mol) | 18 kDa |
Name | IL-33 |
Quantity | 10μg |
Description
The Recombinant Human IL-33 Protein is derived from E. coli. The Recombinant Human IL-33 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
18 kDa | |
10μg | |
E. coli-derived human IL-33 protein Ser112-Thr270 |
90865 | |
IL-33 | |
Yes |