Learn More
R&D Systems™ Human IL-3 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 203-IL-010
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469403
  • £223.44 / 10µg
Estimated Shipment date
from Supplier 21-02-2020
Description
Description
The Recombinant Human IL-3 Protein is derived from E. coli. The Recombinant Human IL-3 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
AAC08706
3562
IL-3
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived Ala20-Phe152, with and without an N-terminal Met
Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Yes