missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-29/IFN-lambda 1 Recombinant Protein

Brand:  R&D Systems™ 1598-IL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459763

  • £335.29 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Gly20-Thr200, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 282618
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
21.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
282618
IL-29/IFN-lambda 1
25μg
Unlabeled
Videos