missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-28B/IFN-lambda 3 Recombinant Protein

Brand:  R&D Systems™ 5259-IL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465364

  • £340.57 / 25µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Arg30-Val200, with the variant of Lys74Arg and a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, NaCl and EDTA.
Entrez Gene ID: 282617
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS, NaCl and EDTA.
20.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
282617
IL-28B/IFN-lambda 3
25μg
Unlabeled
Videos