missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-26/AK155 Monomer Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1375-IL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449253

  • £420.42 / 25µg

Estimated Shipment date
from Supplier 25-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IL-26/AK155 Monomer Protein is derived from E. coli. The Recombinant Human IL-26/AK155 Monomer Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol.
17.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Monomer and a small amount of disulfide-linked homodimer (no more than 7%)
Yes
55801
IL-26/AK155 Monomer
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Unlabeled
Videos
Documents