missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-24 Recombinant Protein

Brand:  R&D Systems™ 1965-IL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460314

  • £245.53 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Met28-Leu206, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 11009
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
19.5 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
11009
IL-24
25μg
Unlabeled
Videos