missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-20 Recombinant Protein

Brand:  R&D Systems™ 1102-IL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458493

  • £253.45 / 25µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Leu25-Glu176, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 50604
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
17.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
50604
IL-20
25μg
Unlabeled
Videos