missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-2 R beta Recombinant Protein

Brand:  R&D Systems™ 224-2B-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460864

  • £497.92 / 25µg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Measured by its ability to inhibit the IL-15-dependent proliferation of MO7e human megakaryocytic leukemic cells. The ED 50 for this effect is 1.0-3.0 µg/mL in the presence of 4.0 ng/mL of recombinant human IL-15.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Observed Molecular Wt: 29kDa and 31kDa, reducing conditions, Predicted Molecular Wt.: 25kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3560
IL-2 R beta
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos