missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-2 ELISpot Kit

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Brand:  R&D Systems™ EL202

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13445095

  • £435.12 / 1 set

Estimated Shipment date
from Supplier 20-12-2019

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control

Description

Description

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
Specifications

Specifications

Diagnostics
Human IL-2 ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
One 96-well PVDF-backed plate coated w/polyclonal Ab for human IL-2, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT chromogen, Positive control
Whole Cells
One 96-well plate
Videos
Documents