missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-2 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
POA
Specifications
Product Type | Human IL-2 ELISpot Kit |
---|---|
For Use With (Application) | Diagnostics |
Sample Type | Whole Cells |
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously. Complete ELISpot kits are ready-to-r
Product Features:- An economical alternative to ELISpot Kits
- Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
- Generic development protocols provide direction
Specifications
Human IL-2 ELISpot Kit | |
Diagnostics | |
Whole Cells |