Learn More
R&D Systems™ Human IL-2 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Brand:  R&D Systems™ XEL202
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443286
  • £2219.94 / Pack of 6
Estimated Shipment
27-06-2019
Description
Description
Product Features:
  • An economical alternative to ELISpot Kits
  • Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
  • Generic development protocols provide direction to start an optimization protocol
  • Customize the assay to your specific needs
Module content:
  • Capture Antibody
  • Biotinylated-conjugated Detection Antibody
Specifications
Specifications
Diagnostics
Human IL-2 ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Six 96-well PVDF-backed plates coated w/polyclonal Ab for human IL-2, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control
Whole Cells
Six 96-well plates
Videos