missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-2 (Cys145Ser) Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1081-IL-020/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428523

  • £255.78 / 20µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IL-2 (Cys145Ser) Protein is derived from E. coli. The Recombinant Human IL-2 (Cys145Ser) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Sodium Acetate.
15.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3558
IL-2 (Cys145Ser)
20μg
6 months from date of receipt, 2 to 8° C as supplied.
Yes
Videos
Documents