missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-1ra/IL-1F3 Recombinant Protein

Brand:  R&D Systems™ 280-RA-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461874

  • £664.29 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3557
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3557
IL-1ra/IL-1F3
50μg
Unlabeled
Videos