missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-17F Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 112744 |
---|---|
Molecular Weight (g/mol) | 15 kDa (monomer) |
Name | IL-17F |
Quantity | 25μg |
Source | E. coli-derived human IL-17F protein Arg31-Gln163,with an N-terminal Met |
Description
The Recombinant Human IL-17F Protein is derived from E. coli. The Recombinant Human IL-17F Protein has been validated for the following applications: Bioactivity.Specifications
112744 | |
IL-17F | |
E. coli-derived human IL-17F protein Arg31-Gln163,with an N-terminal Met | |
Disulfide-linked homodimer | |
Recombinant |
15 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated |