missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-17D Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 53342 |
Molecular Weight (g/mol) | 20 kDa (monomer) |
Name | IL-17D |
Quantity | 25μg |
Description
The Recombinant Human IL-17D Protein is derived from E. coli. The Recombinant Human IL-17D Protein has been validated for the following applications: Binding Activity.Specifications
Unconjugated | |
20 kDa (monomer) | |
25μg | |
E. coli-derived human IL-17D protein Ala18-Pro202 |
53342 | |
IL-17D | |
Yes |