missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-17B Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1248-IB-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458973

  • £290.08 / 25µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IL-17B Protein is derived from E. coli. The Recombinant Human IL-17B Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein.
18.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human IL-17B protein Gln21-Phe180,with an N-terminal Met
Unconjugated
27190
IL-17B
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents