Learn More
R&D Systems™ Human IL-17 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Brand:  R&D Systems™ XEL317
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443296
  • £2610.72 / Pack of 6
Call for latest availability
+44 (0)1509 555 500
Description
Description
This kit also recognizes primate IL-17
Specifications
Specifications
Human
Human/Primate IL-17/IL-17A Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Human/Primate IL-17/IL-17A Positive Control
Whole cells
6 x 96-well plates
Quantitative determination of the frequency of cells releasing human IL-17
Human IL-17 ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A