missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-16 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 3603 |
Molecular Weight (g/mol) | 13.3 kDa |
Name | IL-16 |
Recombinant | Yes |
Description
The Recombinant Human IL-16 Protein is derived from E. coli. The Recombinant Human IL-16 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
13.3 kDa | |
Yes |
3603 | |
IL-16 | |
E. coli-derived human IL-16 protein Pro2-Ser130 |