missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-12 Recombinant Protein

Brand:  R&D Systems™ 219-IL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460784

  • £1016.00 / 25µg


Estimated Shipment
27-06-2018
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ile23-Ser328 (p40) & Arg23-Ser219 (p35)
Purity: 97%, by SDS-PAGE with silver staining.
Molecular Weight: 34.7 kDa (p40) & 22.5 kDa (p35)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3592
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
34.7 kDa (p40) & 22.5 kDa (p35)
97%, by SDS-PAGE with silver staining.
Human
Unlabeled
3592
IL-12
25μg
Disulfide-linked heterodimer
Yes
Videos