missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-11 Recombinant Protein

Brand:  R&D Systems™ 218-IL-005/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460754

  • £279.85 / 5µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Pro22-Leu199
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS and EDTA.
Entrez Gene ID: 3589
Regulatory Status: RUO

Specifications

Specifications

Supplied as a 0.2μm filtered solution in PBS and EDTA.
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3589
IL-11
5μg
Unlabeled
Videos