missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-11 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 218-IL-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13450754

  • £311.64 / 5µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IL-11 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-11 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin
Yes
3589
IL-11
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents