Learn More
R&D Systems™ Human IL-10 Recombinant Protein, (aa 19-178)
Brand: R&D Systems™ 1064-IL-010
Code : Z1
Additional Details : Weight : 0.00010kg
Description
Source: E. coli-derived, Ser19-Asn178, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3586
Regulatory Status: RUO
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
18.8 kDa | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. | |
Human | |
Unlabeled |
3586 | |
IL-10 | |
10μg | |
Noncovalently-linked homodimer | |
Yes |