missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-1 beta/IL-1F2 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 201-LB-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449373

  • £1004.01 / 25µg

Estimated Shipment date
from Supplier 26-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Human IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in PBS.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3553
IL-1 beta/IL-1F2
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Yes
Videos
Documents