missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-1 alpha/IL-1F1 Recombinant Protein

Brand:  R&D Systems™ 200-LA-002

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460394

  • £201.96 / 2µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Ser113-Ala271
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3552
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3552
IL-1 alpha/IL-1F1
2μg
Unlabeled
Videos